Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trichloronicotinic acid 97%

Supplier: AMBEED, INC
Warning

A147092-1G A147092-250MG
77247-150EA 23 USD InStock
77247-150 77247-152
2,4,6-Trichloronicotinic acid 97%
2,4,6-Trichloronicotinic acid
Precaución: for Research Use Only


Formula: C₆H₂Cl₃NO₂
MW: 226.44 g/mol
MDL Number: MFCD09038102
CAS Number: 69422-72-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty