Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(6'-(2-Hydroxy-2-methylpropoxy)-4-((5-methylpyridin-3-yl)oxy)-[3,3'-bipyridin]-6-yl)-3-methylurea 98%

Supplier: AMBEED, INC
Warning

A147455-10MG A147455-1MG A147455-25MG A147455-50MG A147455-5MG
77259-090EA 56 USD InStock
77259-090 77259-092 77259-094 77259-096 77259-098
1-(6'-(2-Hydroxy-2-methylpropoxy)-4-((5-methylpyridin-3-yl)oxy)-[3,3'-bipyridin]-6-yl)-3-methylurea 98%
1-(6'-(2-Hydroxy-2-methylpropoxy)-4-((5-methylpyridin-3-yl)oxy)-[3,3'-bipyridin]-6-yl)-3-methylurea
Precaución: for Research Use Only


Formula: C₂₂H₂₅N₅O₄
MDL Number: MFCD30533320
CAS Number: 1442684-77-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty