Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(1-Benzylpiperidin-4-yl)-4-iodobenzamide 98%

Supplier: AMBEED, INC
Warning

A147803-100MG A147803-10MG A147803-1MG A147803-50MG A147803-5MG
77235-262EA 75 USD InStock
77235-262 77235-264 77235-266 77235-268 77235-270
N-(1-Benzylpiperidin-4-yl)-4-iodobenzamide 98%
N-(1-Benzylpiperidin-4-yl)-4-iodobenzamide
Precaución: for Research Use Only


Formula: C₁₉H₂₁IN₂O
MDL Number: MFCD00673910
CAS Number: 155798-08-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty