Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-9,9-dimethyl-9H-fluorene 97%

Supplier: AMBEED, INC
Warning

A148004-1G A148004-250MG A148004-5G A148004-25G
77235-288EA 30 USD InStock
77235-288 77235-290 77235-292 77746-035
3-Bromo-9,9-dimethyl-9H-fluorene 97%
3-Bromo-9,9-dimethyl-9H-fluorene
Precaución: for Research Use Only


Formula: C₁₅H₁₃Br
MDL Number: MFCD28955974
CAS Number: 1190360-23-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty