Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Carbethoxyimidazole 97%

Supplier: AMBEED, INC
Warning

A148579-100G A148579-10G A148579-1G A148579-25G A148579-5G
77235-320EA 319 USD InStock
77235-320 77235-322 77235-324 77235-326 77235-328
1-Carbethoxyimidazole 97%
1-Carbethoxyimidazole
Precaución: for Research Use Only


Formula: C₆H₈N₂O₂
MDL Number: MFCD00014498
CAS Number: 19213-72-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty