Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Butylphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A148616-100G A148616-10G A148616-1G A148616-25G A148616-5G
77272-872EA 149 USD InStock
77272-872 77272-874 77272-876 77272-878 77272-880
4-Butylphenylboronic acid 98%
4-Butylphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₅BO₂
MDL Number: MFCD02093926
CAS Number: 145240-28-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty