Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-O-Benzyl-4-(hydroxymethyl-1,2-O-isopropylidene)-α-D-erythropentofuranose 95%

Supplier: AMBEED, INC
Warning

A148765-10G A148765-1G A148765-25G
77286-054EA 31 USD InStock
77286-054 77286-056 77748-035
3-O-Benzyl-4-(hydroxymethyl-1,2-O-isopropylidene)-α-D-erythropentofuranose 95%
3-O-Benzyl-4-(hydroxymethyl-1,2-O-isopropylidene)-α-D-erythropentofuranose
Precaución: for Research Use Only


Formula: C₁₆H₂₂O₆
MW: 310.342 g/mol
MDL Number: MFCD08704144
CAS Number: 63593-03-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty