Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-n-Pentylcinnamoyl)anthranilic acid 98%

Supplier: AMBEED, INC
Warning

A148872-100MG A148872-1MG A148872-25MG A148872-50MG
77235-342EA 152 USD InStock
77235-342 77235-344 77235-346 77235-348
N-(4-n-Pentylcinnamoyl)anthranilic acid 98%
N-(4-n-Pentylcinnamoyl)anthranilic acid
Precaución: for Research Use Only


Formula: C₂₁H₂₃NO₃
MW: 337.419 g/mol
MDL Number: MFCD00211365
CAS Number: 110683-10-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty