Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H-Asp(OtBu)-OMe·HCl 95%

Supplier: AMBEED, INC
Warning

A149317-10G A149317-1G A149317-25G A149317-5G
77259-160EA 46 USD InStock
77259-160 77259-162 77259-164 77259-166
H-Asp(OtBu)-OMe·HCl 95%
H-Asp(OtBu)-OMe·HCl
Precaución: for Research Use Only


Formula: C₉H₁₈ClNO₄
MDL Number: MFCD00153427
CAS Number: 2673-19-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty