Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Aminomethyl)phenol 97%

Supplier: AMBEED, INC
Warning

A149449-10G A149449-1G A149449-250MG A149449-25G A149449-5G
77272-924EA 50 USD InStock
77272-924 77272-926 77272-928 77272-930 77272-932
3-(Aminomethyl)phenol 97%
3-(Aminomethyl)phenol
Precaución: for Research Use Only


Formula: C₇H₉NO
MW: 123.155 g/mol
MDL Number: MFCD00798977
CAS Number: 73604-31-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty