Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-N-amido-PEG1-acid 97%

Supplier: AMBEED, INC
Warning

A150060-100MG A150060-1G A150060-250MG
77247-202EA 16 USD InStock
77247-202 77247-204 77247-206
Fmoc-N-amido-PEG1-acid 97%
Fmoc-N-amido-PEG1-acid
Precaución: for Research Use Only


Formula: C₂₀H₂₁NO₅
MDL Number: MFCD16294357
CAS Number: 1654740-73-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty