Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Brigatinib (AP26113) ≥98%

Supplier: AMBEED, INC
Warning

A151843-10MG A151843-1MG A151843-2MG A151843-5MG A151843-1G
77235-460EA 11 USD InStock
77235-460 77235-462 77235-464 77235-466 77746-050
Brigatinib (AP26113) ≥98%
Brigatinib (AP26113)
Precaución: for Research Use Only


Formula: C₂₉H₃₉ClN₇O₂P
MW: 584.09 g/mol
MDL Number: MFCD29472221
CAS Number: 1197953-54-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty