Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-1-naphthaldehyde 98%

Supplier: AMBEED, INC
Warning

A152589-100G A152589-10G A152589-25G A152589-500G A152589-5G
77259-300EA 80 USD InStock
77259-300 77259-302 77259-304 77259-306 77259-308
2-Methoxy-1-naphthaldehyde 98%
2-Methoxy-1-naphthaldehyde
Precaución: for Research Use Only


Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 339
MDL Number: MFCD00004004
CAS Number: 5392-12-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty