Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 1-benzyl-1H-indole-5-carboxylate 97%

Supplier: AMBEED, INC
Warning

A152695-100MG A152695-1G A152695-250MG
77259-320EA 150 USD InStock
77259-320 77259-322 77259-324
Methyl 1-benzyl-1H-indole-5-carboxylate 97%
Methyl 1-benzyl-1H-indole-5-carboxylate
Precaución: for Research Use Only


Formula: C₁₇H₁₅NO₂
MDL Number: MFCD26394992
CAS Number: 192997-32-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty