Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-chloro-5-iodobenzoate 98%

Supplier: AMBEED, INC
Warning

A152881-100MG A152881-10G A152881-1G A152881-250MG A152881-25G A152881-5G
77259-330EA 5 USD InStock
77259-330 77259-332 77259-334 77259-336 77748-050 77748-051
Methyl-2-chloro-5-iodobenzoate 98%
Methyl 2-chloro-5-iodobenzoate
Precaución: for Research Use Only


Formula: C₈H₆ClIO₂
MW: 296.4900 g/mol
MDL Number: MFCD07779253
CAS Number: 620621-48-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty