Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-((1R,2S)-2-(4-Allyl-2,6-dimethoxyphenoxy)-1-hydroxypropyl)-2-methoxyphenol 98%

Supplier: AMBEED, INC
Warning

A153206-10MG A153206-1MG A153206-25MG A153206-5MG
77247-350EA 72 USD InStock
77247-350 77247-352 77247-354 77247-356
4-((1R,2S)-2-(4-Allyl-2,6-dimethoxyphenoxy)-1-hydroxypropyl)-2-methoxyphenol 98%
4-((1R,2S)-2-(4-Allyl-2,6-dimethoxyphenoxy)-1-hydroxypropyl)-2-methoxyphenol
Precaución: for Research Use Only


Formula: C₂₁H₂₆O₆
MDL Number: MFCD13195532
CAS Number: 171485-39-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty