Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-nitrobenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A153734-10G A153734-5G
77286-206EA 31 USD InStock
77286-206 77286-208
4-Chloro-2-nitrobenzaldehyde 98%
4-Chloro-2-nitrobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
MDL Number: MFCD00056104
CAS Number: 5551-11-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty