Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-3-bromopiperidine 97%

Supplier: AMBEED, INC
Warning

A154878-100MG A154878-1G A154878-250MG A154878-5G
77235-588EA 7 USD InStock
77235-588 77235-590 77235-592 77549-964
1-Boc-3-bromopiperidine 97%
1-Boc-3-bromopiperidine
Precaución: for Research Use Only


Formula: C₁₀H₁₈BrNO₂
MDL Number: MFCD07779077
CAS Number: 849928-26-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty