Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dichloropicolinic acid 98%

Supplier: AMBEED, INC
Warning

A154914-100G A154914-10G A154914-1G A154914-250MG A154914-25G A154914-5G
77286-230EA 235 USD InStock
77286-230 77286-232 77286-234 77286-236 77286-238 77286-240
4,6-Dichloropicolinic acid 98%
4,6-Dichloropicolinic acid
Precaución: for Research Use Only


Formula: C₆H₃Cl₂NO₂
MW: 192.0000 g/mol
MDL Number: MFCD09910267
CAS Number: 88912-25-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty