Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluoroaniline 97%

Supplier: AMBEED, INC
Warning

A155079-100G A155079-500G A155079-25G
77286-250EA 41 USD InStock
77286-250 77286-252 77300-104
4-Bromo-2-fluoroaniline 97%
4-Bromo-2-fluoroaniline
Precaución: for Research Use Only


Formula: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD00010221
CAS Number: 367-24-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty