Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-fluorobenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A155319-100G A155319-1G A155319-25G A155319-5G
77273-114EA 293.99 USD InStock
77273-114 77273-116 77273-118 77273-120
3-Bromo-5-fluorobenzyl alcohol 98%
3-Bromo-5-fluorobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₆BrFO
MDL Number: MFCD04116320
CAS Number: 216755-56-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty