Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-6-nitrophenol 98%

Supplier: AMBEED, INC
Danger

A155664-100G A155664-1G A155664-25G A155664-500G A155664-5G
77247-450EA 135 USD InStock
77247-450 77247-452 77247-454 77247-456 77247-458
2,4-Dichloro-6-nitrophenol 98%
2,4-Dichloro-6-nitrophenol
Precaución: for Research Use Only


Formula: C₆H₃Cl₂NO₃
MDL Number: MFCD00007101
CAS Number: 609-89-2
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty