Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2-nitroaniline 98%

Supplier: AMBEED, INC
Warning

A156514-100G A156514-10G A156514-25G A156514-500G A156514-5G
77273-138EA 23 USD InStock
77273-138 77273-140 77273-142 77273-144 77273-146
5-Fluoro-2-nitroaniline 98%
5-Fluoro-2-nitroaniline
Precaución: for Research Use Only


Formula: C₆H₅FN₂O₂
MW: 156.12
MDL Number: MFCD00034065
CAS Number: 2369-11-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty