Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitro-o-toluonitrile 98%

Supplier: AMBEED, INC
Warning

A158415-10G A158415-1G A158415-25G A158415-5G
77273-216EA 49 USD InStock
77273-216 77273-218 77273-220 77273-222
4-Nitro-o-toluonitrile 98%
4-Nitro-o-toluonitrile
Precaución: for Research Use Only


Formula: C₈H₆N₂O₂
MW: 162.15
MDL Number: MFCD03095378
CAS Number: 89001-53-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty