Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-methylbenzoic acid 97%

Supplier: AMBEED, INC
Warning

A158504-5G A158504-25G
77259-500EA 12 USD InStock
77259-500 77748-072
2-Chloro-4-methylbenzoic acid 97%
2-Chloro-4-methylbenzoic acid


Formula: C₈H₇ClO₂
MDL Number: MFCD03094667
CAS Number: 7697-25-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty