Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

OTSSP167 99%

Supplier: AMBEED, INC
Warning

A158718-10MG A158718-1MG A158718-2MG A158718-50MG A158718-5MG
77235-750EA 124 USD InStock
77235-750 77235-752 77235-754 77235-756 77235-758
OTSSP167 99%
OTSSP167
Precaución: for Research Use Only


Formula: C₂₅H₂₈Cl₂N₄O₂
MW: 487.42 g/mol
MDL Number: MFCD23160047
CAS Number: 1431697-89-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty