Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-1H-indole-3-carbaldehyde 98%

Supplier: AMBEED, INC
Warning

A158826-1G A158826-250MG A158826-5G
77286-370EA 29 USD InStock
77286-370 77286-372 77286-374
4-Fluoro-1H-indole-3-carbaldehyde 98%
4-Fluoro-1H-indole-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₉H₆FNO
MW: 163.148 g/mol
MDL Number: MFCD00056930
CAS Number: 23073-31-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty