Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxyisophthalic acid 97%

Supplier: AMBEED, INC
Warning

A158992-100G A158992-10G A158992-1G A158992-5G A158992-25G
77259-514EA 627 USD InStock
77259-514 77259-516 77259-518 77259-520 77259-522 77748-079
5-Methoxyisophthalic acid 97%
5-Methoxyisophthalic acid
Precaución: for Research Use Only


Formula: C₉H₈O₅
MDL Number: MFCD00272364
CAS Number: 46331-50-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty