Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4,6-dimethylbenzenol 96%

Supplier: AMBEED, INC
Warning

A159140-10G A159140-1G A159140-250MG A159140-5G
77259-534EA 105 USD InStock
77259-534 77259-536 77259-538 77259-540
2-Bromo-4,6-dimethylbenzenol 96%
2-Bromo-4,6-dimethylbenzenol
Precaución: for Research Use Only


Formula: C₈H₉BrO
MDL Number: MFCD08443978
CAS Number: 15191-36-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty