Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-fluorophenol 98%

Supplier: AMBEED, INC
Warning

A159557-100G A159557-10G A159557-25G A159557-500G A159557-5G
77286-406EA 175 USD InStock
77286-406 77286-408 77286-410 77286-412 77286-414
4-Bromo-3-fluorophenol 98%
4-Bromo-3-fluorophenol
Precaución: for Research Use Only


Formula: C₆H₄BrFO
MW: 191
MDL Number: MFCD00051907
CAS Number: 121219-03-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty