Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-Chloro-N-(2-oxo-2-((1-phenylethyl)amino)ethyl)benzamide 98%

Supplier: AMBEED, INC
Warning

A159562-10MG A159562-1MG A159562-50MG A159562-5MG
77235-802EA 39 USD InStock
77235-802 77235-804 77235-806 77235-808
(S)-3-Chloro-N-(2-oxo-2-((1-phenylethyl)amino)ethyl)benzamide 98%
(S)-3-Chloro-N-(2-oxo-2-((1-phenylethyl)amino)ethyl)benzamide
Precaución: for Research Use Only


Formula: C₁₇H₁₇ClN₂O₂
MDL Number: MFCD29924732
CAS Number: 1802326-66-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty