Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Benzyloxy)propan-1-amine 97%

Supplier: AMBEED, INC
Warning

A159567-100MG A159567-10G A159567-1G A159567-250MG A159567-5G
77273-250EA 50 USD InStock
77273-250 77273-252 77273-254 77273-256 77273-258
3-(Benzyloxy)propan-1-amine 97%
3-(Benzyloxy)propan-1-amine
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
MDL Number: MFCD07339550
CAS Number: 16728-64-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty