Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopropylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A160037-100G A160037-10G A160037-1G A160037-25G A160037-5G
77259-562EA 166 USD InStock
77259-562 77259-564 77259-566 77259-568 77259-570
Isopropylboronic acid pinacol ester 98%
Isopropylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₉H₁₉BO₂
MDL Number: MFCD05663856
CAS Number: 76347-13-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty