Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromoacetamide 96%

Supplier: AMBEED, INC
Danger

A161414-1G A161414-25G A161414-5G A161414-100G
77259-618EA 8 USD InStock
77259-618 77259-620 77259-622 77746-103
Bromoacetamide 96%
Bromoacetamide
Precaución: for Research Use Only


Formula: BrCH₂CONH₂
MDL Number: MFCD00008025
CAS Number: 683-57-8
UN: 2923
ADR: 8,6.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty