Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-3-Chloro-1-phenyl-1-propanol 99%

Supplier: AMBEED, INC
Warning

A161565-1G A161565-250MG A161565-25G A161565-10G A161565-5G
77247-668EA 6 USD InStock
77247-668 77247-670 77247-672 77746-104 77746-105
(R)-(+)-3-Chloro-1-phenyl-1-propanol 99%
(R)-(+)-3-Chloro-1-phenyl-1-propanol
Precaución: for Research Use Only


Formula: C₉H₁₁ClO
MW: 170.64
MDL Number: MFCD00075128
CAS Number: 100306-33-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty