Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine 98%

Supplier: AMBEED, INC
Warning

A161611-100MG A161611-250MG
77247-674EA 13 USD InStock
77247-674 77247-676
(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine 98%
(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine
Precaución: for Research Use Only


Formula: C₃₆H₄₄FeP₂
MDL Number: MFCD00800281
CAS Number: 162291-01-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty