Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-nitro-1H-indazole 97%

Supplier: AMBEED, INC
Warning

A162433-10G A162433-1G A162433-250MG A162433-5G
77273-378EA 46 USD InStock
77273-378 77273-380 77273-382 77273-384
3-Chloro-5-nitro-1H-indazole 97%
3-Chloro-5-nitro-1H-indazole
Precaución: for Research Use Only


Formula: C₇H₄ClN₃O₂
MDL Number: MFCD00005692
CAS Number: 4812-45-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty