Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrazine-2,3-dicarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A164253-100G A164253-25G A164253-500G A164253-5G A164253-1000G
77286-548EA 46 USD InStock
77286-548 77286-550 77286-552 77286-554 77286-556 77748-097
Pyrazine-2,3-dicarboxylic acid 98%
Pyrazine-2,3-dicarboxylic acid
Precaución: for Research Use Only


Formula: C₆H₄N₂O₄
MW: 168.11 g/mol
MDL Number: MFCD00006131
CAS Number: 89-01-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty