Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromobenzaldehyde ethylene acetal 98% (by GC)

Supplier: AMBEED, INC
Warning

A164430-100G A164430-10G A164430-1G A164430-25G A164430-5G
77259-742EA 59 USD InStock
77259-742 77259-744 77259-746 77259-748 77259-750
2-Bromobenzaldehyde ethylene acetal 98% (by GC)
2-Bromobenzaldehyde ethylene acetal
Precaución: for Research Use Only


Formula: C₉H₉BrO₂
MDL Number: MFCD00155124
CAS Number: 34824-58-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty