Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Piperidin-4-yl)phenol hydrobromide 97%

Supplier: AMBEED, INC
Warning

A164679-10G A164679-1G A164679-250MG A164679-5G
77259-752EA 67 USD InStock
77259-752 77259-754 77259-756 77259-758
4-(Piperidin-4-yl)phenol hydrobromide 97%
4-(Piperidin-4-yl)phenol hydrobromide
Precaución: for Research Use Only


Formula: C₁₁H₁₆BrNO
MDL Number: MFCD30607288
CAS Number: 1869912-48-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty