Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indole-2-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A164849-100G A164849-10G A164849-25G A164849-5G A164849-1000G
77273-492EA 45 USD InStock
77273-492 77273-494 77273-496 77273-498 77748-099
Indole-2-carboxylic acid 98%
Indole-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₇NO₂
MW: 161.16 g/mol
MDL Number: MFCD00005611
CAS Number: 1477-50-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty