Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium ((2R,3S,4R,5R)-3,4-dihydroxy-5-(6-hydroxy-9H-purin-9-yl)tetrahydrofuran-2-yl)methyl phosphate heptahydrate 98%

Supplier: AMBEED, INC
Warning

A165095-100G A165095-10G A165095-25G A165095-5G
77247-768EA 105 USD InStock
77247-768 77247-770 77247-772 77247-774 77247-776
Sodium ((2R,3S,4R,5R)-3,4-dihydroxy-5-(6-hydroxy-9H-purin-9-yl)tetrahydrofuran-2-yl)methyl phosphate heptahydrate 98%
Sodium ((2R,3S,4R,5R)-3,4-dihydroxy-5-(6-hydroxy-9H-purin-9-yl)tetrahydrofuran-2-yl)methyl phosphate heptahydrate
Precaución: for Research Use Only


Formula: C₁₀H₂₅N₄Na₂O₁₅P
MW: 518.27 g/mol
MDL Number: MFCD30536239
CAS Number: 849725-39-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty