Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dimethylisonicotinaldehyde 98%

Supplier: AMBEED, INC
Warning

A165215-100MG A165215-1G A165215-250MG A165215-5G
77247-782EA 20 USD InStock
77247-782 77247-784 77247-786 77247-788
2,6-Dimethylisonicotinaldehyde 98%
2,6-Dimethylisonicotinaldehyde
Precaución: for Research Use Only


Formula: C₈H₉NO
MDL Number: MFCD06410687
CAS Number: 18206-06-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty