Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrobenzotriazole 97%

Supplier: AMBEED, INC
Danger

A165821-100MG A165821-250MG
77286-608EA 11 USD InStock
77286-608 77286-610
4-Nitrobenzotriazole 97%
4-Nitrobenzotriazole
Precaución: for Research Use Only


Formula: C₆H₄N₄O₂
MDL Number: MFCD00160033
CAS Number: 6299-39-4
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty