Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Fluorophenyl)acetone 97%

Supplier: AMBEED, INC
Warning

A166150-1G A166150-250MG A166150-25G
77273-548EA 57 USD InStock
77273-548 77273-550 77273-552
(3-Fluorophenyl)acetone 97%
(3-Fluorophenyl)acetone
Precaución: for Research Use Only


Formula: C₉H₉FO
MDL Number: MFCD00061144
CAS Number: 1737-19-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty