Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-iodo-5-methylpyridine 97%

Supplier: AMBEED, INC
Warning

A166263-1G A166263-250MG A166263-5G
77247-836EA 21 USD InStock
77247-836 77247-838 77247-840
2-Amino-3-iodo-5-methylpyridine 97%
2-Amino-3-iodo-5-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₇IN₂
MDL Number: MFCD09037445
CAS Number: 211308-79-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty