Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-5-methylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A166372-10G A166372-1G A166372-250MG A166372-25G A166372-5G
77273-558EA 160 USD InStock
77273-558 77273-560 77273-562 77273-564 77273-566
3-Hydroxy-5-methylbenzoic acid 98%
3-Hydroxy-5-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₈H₈O₃
MW: 152.147 g/mol
MDL Number: MFCD09833213
CAS Number: 585-81-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty