Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-bromo-3-phenylpropanoate 95%

Supplier: AMBEED, INC
Danger

A166436-100MG A166436-1G A166436-250MG A166436-5G
77259-786EA 38 USD InStock
77259-786 77259-788 77259-790 77259-792
Methyl 2-bromo-3-phenylpropanoate 95%
Methyl 2-bromo-3-phenylpropanoate
Precaución: for Research Use Only


Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD07786597
CAS Number: 3196-22-3
UN: 1760
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty