Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-chloro-5-fluorobenzene 98%

Supplier: AMBEED, INC
Warning

A167274-100G A167274-10G A167274-500G A167274-5G A167274-25G
77286-644EA 38 USD InStock
77286-644 77286-646 77286-648 77286-650 77300-884
1-Bromo-3-chloro-5-fluorobenzene 98%
1-Bromo-3-chloro-5-fluorobenzene
Precaución: for Research Use Only


Formula: C₆H₃BrClF
MW: 209.44
MDL Number: MFCD00070747
CAS Number: 33863-76-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty