Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-D-Tic-OH 98%

Supplier: AMBEED, INC
Warning

A167918-100G A167918-10G A167918-1G A167918-250MG A167918-5G
77273-600EA 171 USD InStock
77273-600 77273-602 77273-604 77273-606 77273-608
Boc-D-Tic-OH 98%
Boc-D-Tic-OH
Precaución: for Research Use Only


Formula: C₁₅H₁₉NO₄
MDL Number: MFCD00143818
CAS Number: 115962-35-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty